

2-Chlorobenzoyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:609-65-4
:
Formula:C₇H₄Cl₂O
MDL Number:MFCD00000660
:175.01 g/mol
Boiling Pt:230°C
Melting Pt:–4 °C
Density:1,38 g/cm³ (20 °C)
Flash Pt:124°C
Storage Temperature:Ambient
EINECS:210-194-0
UN:3265
ADR:8,II
:GHS05
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Argentmetric Titration):min. 98.0 %
Specification Test Results
Frequently Bought Together