Order Entry
Italy
ContactUsLinkComponent
2,7-Diisopropylnaphthalene ≥97.0% (by GC)
2,7-Diisopropylnaphthalene ≥97.0% (by GC)
Catalog # TCIAD2422-25ML
Supplier:  TCI
CAS Number:  
undefined
2,7-Diisopropylnaphthalene ≥97.0% (by GC)
Catalog # TCIAD2422-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
Boiling Pt:280°C
 :212.33 g/mol
EINECS:254-929-3
CAS Number:40458-98-8
Formula:C₁₆H₂₀
MDL Number:MFCD00671522