
2,6-Difluorobenzamide ≥97.0% (by total nitrogen basis)
Catalog # TCIAD1907-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Melting point:144.0 to 147.0 deg-C
Specification Test Results
About this item
:
:157.12 g/mol
Boiling Pt:284 °C (1013 hPa)
Melting Pt:144…145.5 °C
EINECS:241-972-8
:GHS07
CAS Number:18063-03-1
Formula:C₇H₅F₂NO
MDL Number:MFCD00007972