2,5-Dimethylindole ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1196-79-8
:
Formula:C₁₀H₁₁N
MDL Number:MFCD00005621
:145.2 g/mol
Boiling Pt:282 °C (1013 hPa)
Melting Pt:116 °C
Density:1,08 g/cm³ (20 °C)
EINECS:214-816-1
Specification Test Results
- Appearance:White to Brown to Dark purple powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:113.0 to 117.0 deg-C
Specification Test Results
Frequently Bought Together