Order Entry
Italy
ContactUsLinkComponent
2,4-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
 
2,4-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
  TCIAD1882-10G
 :  TCI
 :  
undefined
 
2,4-Difluorobenzoyl chloride ≥98.0% (by GC, titration analysis)
  TCIAD1882-10G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %

 

 

 :
 :176.55 g/mol
 :70°C
 :1,42 g/cm³ (20 °C)
 :82°C
 :276-690-4
 :3265
 :8,III
 :GHS05
 :72482-64-5
 :C₇H₃ClF₂O
 :MFCD00000658