Order Entry
Italy
ContactUsLinkComponent
2,4-Dibromopentane ≥98.0% (by GC)
Flammable
Irritant
2,4-Dibromopentane ≥98.0% (by GC)
Catalog # TCIAD1410-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2,4-Dibromopentane ≥98.0% (by GC)
Catalog # TCIAD1410-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 % (total of isomers)

Specification Test Results

About this item

 :
 :229.94 g/mol
Boiling Pt:40°C
EINECS:243-031-7
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:19398-53-9
Formula:H₃CBrCHCH₂BrCHCH₃
MDL Number:MFCD00039176