

2,3-Diaminonaphthalene ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:771-97-1
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004116
:158.2 g/mol
Boiling Pt:343 °C (1013 hPa)
Melting Pt:199 °C
Density:1,0968 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:212-241-0
:GHS07
:GHS08
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystaline
- Purity (GC):min. 97.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Specification Test Results
Related Information
Frequently Bought Together