Order Entry
Italy
ContactUsLinkComponent
2,2-Diphenylpropionic acid ≥96.0% (by HPLC, titration analysis)
Irritant
2,2-Diphenylpropionic acid ≥96.0% (by HPLC, titration analysis)
Catalog # TCIAD1778-5G
Supplier:  TCI
CAS Number:  
undefined
Irritant
2,2-Diphenylpropionic acid ≥96.0% (by HPLC, titration analysis)
Catalog # TCIAD1778-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 96.0 area%
  • Purity (Neutralization titration):
    min. 96.0 %
  • Melting point:
    173.0 to 177.0 deg-C

Specification Test Results

About this item

 :
 :226.27 g/mol
Boiling Pt:300 °C (1013 hPa)
Melting Pt:178 °C
EINECS:226-924-6
 :GHS07
CAS Number:5558-66-7
Formula:C₁₅H₁₄O₂
MDL Number:MFCD00004189