Order Entry
Italy
ContactUsLinkComponent
2,2-Dimethylvaleric acid ≥98.0% (by GC, titration analysis)
Corrosivo
2,2-Dimethylvaleric acid ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI
Certificati
undefined
Corrosivo
2,2-Dimethylvaleric acid ≥98.0% (by GC, titration analysis)
Nome del fornitore:  TCI

Risultati dei test sulle specifiche

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 98.0 %
  • Purity (Neutralization titration):
    min. 98.0 %

Risultati dei test sulle specifiche

Frequently Bought Together