



2,2,3-Trimethylpentane ≥99.0% (by GC)
Catalog # TCIAT0689-1ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- Seq:2070
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
About this item
:
:114.23 g/mol
Boiling Pt:110°C
Melting Pt:–112.2 °C
Density:0,7161 g/cm³ (20 °C)
Flash Pt:8°C
Storage Temperature:Ambient
EINECS:209-266-4
UN:1262
ADR:3,II
:GHS02
:GHS07
:GHS08
:GHS09
CAS Number:564-02-3
Formula:C₈H₁₈
MDL Number:MFCD00059241