

2,4,6-Trichlorophenyl isocyanate ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2505-31-9
:
Formula:C₇H₂Cl₃NO
MDL Number:MFCD00013845
Boiling Pt:101°C
Melting Pt:66…67 °C
UN:2206
ADR:6.1,III
:222.46 g/mol
:GHS06
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to Light yellow to Light orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:65.0 to 69.0 deg-C
Specification Test Results
Frequently Bought Together