p-Cresol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: 1-Hydroxy-4-methylbenzene, 4-Methylphenol, 4-Hydroxytoluene
CAS Number:106-44-5
:
Formula:C₇H₈O
MDL Number:MFCD00002376
:108.14 g/mol
Merck Index:14,02579
Boiling Pt:202°C
Melting Pt:32…34 °C
Density:1,034 g/cm³ (20 °C)
Flash Pt:86 °C
Storage Temperature:Ambient
EINECS:203-398-6
UN:3455
ADR:6.1,II
:GHS05
:GHS06
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Light yellow powder to lump
- Purity (GC):min. 99.0 %
- Solubility in Sodium hydroxide solution:almost transparency
Specification Test Results
Frequently Bought Together