
5-Methoxy-m-cresol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3209-13-0
:
Formula:C₈H₁₀O₂
MDL Number:MFCD00059261
:138.17 g/mol
EINECS:221-716-1
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:62.0 to 64.0 deg-C
Specification Test Results
Frequently Bought Together