
5-Bromo-α,α,α-trifluoro-2-nitrotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:344-38-7
:
Formula:C₇H₃BrF₃NO₂
MDL Number:MFCD00014706
:270.01 g/mol
Boiling Pt:100°C
Melting Pt:40…45 °C
Density:1,799 g/cm³ (25 °C)
:GHS07
Specification Test Results
- Appearance:Light orange to Yellow to Green powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:32.0 to 38.0 deg-C
Specification Test Results
Frequently Bought Together