
4-Bromo-m-cresol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:14472-14-1
:
Formula:C₇H₇BrO
MDL Number:MFCD00079723
:187.04 g/mol
Boiling Pt:145°C
Melting Pt:63 °C
EINECS:238-464-3
:GHS07
Specification Test Results
- Appearance:White to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:66.0 to 70.0 deg-C
Specification Test Results
Frequently Bought Together