

2-Bromophenyl isothiocyanate ≥97.0% (by GC)
Catalog # TCIAI0523-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Purity (GC):min. 97.0 %
Specification Test Results
About this item
:
:214.09 g/mol
Boiling Pt:118°C
Melting Pt:20 °C
Density:1,59 g/cm³ (20 °C)
UN:1760
ADR:8,III
:GHS05
:GHS07
CAS Number:13037-60-0
Formula:C₇H₄BrNS
MDL Number:MFCD00004799