


3-Nitro-p-toluidine ≥98.0% (by titrimetric analysis)
Catalog # TCIAN0278-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:76.0 to 80.0 deg-C
Specification Test Results
About this item
:
:152.15 g/mol
Boiling Pt:306 °C (1013 hPa)
Melting Pt:77…78 °C
Density:1,312 g/cm³ (20 °C)
EINECS:204-314-0
UN:2660
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:119-32-4
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007910