


p-Anisidina ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:104-94-9
:
Formula:C₇H₉NO
MDL Number:MFCD00007864
:123.15 g/mol
Merck Index:14,00667
Boiling Pt:243 °C (1013 hPa)
Melting Pt:55…58 °C
Density:1,2 g/cm³ (20 °C)
Flash Pt:122 °C
Storage Temperature:Refrigerata
EINECS:203-254-2
UN:2811
ADR:6.1,III
:GHS06
:GHS08
:GHS09
Specification Test Results
- Appearance:White to Orange to Green powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:57.0 to 61.0 deg-C
- Solubility in Methanol:very faint turbidity
Specification Test Results
Frequently Bought Together