Order Entry
Italy
ContactUsLinkComponent
 
 
4-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
4-Fluoro-m-toluidine ≥98.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Gray powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    33.0 to 37.0 deg-C

 

Frequently Bought Together