
4-Chloro-3-(trifluoromethyl)aniline ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:320-51-4
:
Formula:C₇H₅ClF₃N
MDL Number:MFCD00007846
:195.57 g/mol
Boiling Pt:130 - 132 °C (27 -rr)
Melting Pt:36…37 °C
Storage Temperature:Ambient
EINECS:206-277-6
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White or Colorless to Yellow to Orange powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Freezing point:34.0 to 37.0 deg-C
Specification Test Results
Frequently Bought Together