

N-Methylethylenediamine ≥97.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:109-81-9
:
Formula:C₃H₁₀N₂
MDL Number:MFCD00008165
:74.13 g/mol
Boiling Pt:116°C
Density:0,84 g/cm³ (20 °C)
Flash Pt:41°C
EINECS:203-707-4
UN:2734
ADR:8,II
:GHS02
:GHS05
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (Neutralization titration):min. 97.0 %
Specification Test Results
Frequently Bought Together