
α,2,6-Trichlorotoluene ≥98.0% (by GC)
Catalog # TCIAD1750-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %
- Melting point:36.0 to 39.0 deg-C
Specification Test Results
About this item
:
:195.47 g/mol
Boiling Pt:119°C
Melting Pt:49…50 °C
EINECS:217-940-4
UN:3261
ADR:8,III
:GHS05
CAS Number:2014-83-7
Formula:C₇H₅Cl₃
MDL Number:MFCD00000897