
α,2,3-Trichlorotoluene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:3290-01-5
:
Formula:C₇H₅Cl₃
MDL Number:MFCD00035817
:195.47 g/mol
Boiling Pt:255°C
EINECS:221-947-8
UN:3265
ADR:8,III
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together