Order Entry
Italy
ContactUsLinkComponent
ω,3,4-Trichloroacetophenone ≥98.0% (by GC, titration analysis)
Corrosive
ω,3,4-Trichloroacetophenone ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1965-5G
Supplier:  TCI
CAS Number:  
undefined
Corrosive
ω,3,4-Trichloroacetophenone ≥98.0% (by GC, titration analysis)
Catalog # TCIAT1965-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Argentmetric Titration):
    min. 98.0 %
  • Melting point:
    42.0 to 45.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

About this item

 :
ADR:8,III
 :223.49 g/mol
UN:1759
 :GHS05
CAS Number:42981-08-8
Formula:C₈H₅Cl₃O
MDL Number:MFCD03970382