Order Entry
Italy
ContactUsLinkComponent
1,8-Nonadiyne ≥97.0% (by GC)
Flammable
Irritant
Health
1,8-Nonadiyne ≥97.0% (by GC)
Catalog # TCIAN0406-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
Health
1,8-Nonadiyne ≥97.0% (by GC)
Catalog # TCIAN0406-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Light yellow to Light orange clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :120.19 g/mol
Boiling Pt:162°C
Melting Pt:–27.3 °C
Density:0,8158 g/cm³ (20 °C)
Flash Pt:41°C
Storage Temperature:Refrigerator
EINECS:219-254-0
UN:3295
ADR:3,III
 :GHS02
 :GHS07
 :GHS08
CAS Number:2396-65-8
Formula:C₉H₁₂
MDL Number:MFCD00008581