Order Entry
Italy
ContactUsLinkComponent
Diethyl pimelate ≥97.0% (by GC)
Diethyl pimelate ≥97.0% (by GC)
Catalog # TCIAP1100-25ML
Supplier:  TCI
CAS Number:  
undefined
Diethyl pimelate ≥97.0% (by GC)
Catalog # TCIAP1100-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :216.28 g/mol
Merck Index:14,07431
Boiling Pt:149°C
Melting Pt:–24 °C
Density:0,99 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:218-083-9
CAS Number:2050-20-6
Formula:C₁₁H₂₀O₄
MDL Number:MFCD00009216