
1,7-Dihydroxynaphthalene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:575-38-2
:
Formula:C₁₀H₈O₂
MDL Number:MFCD00035720
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:175…177 °C
Storage Temperature:Ambient
EINECS:209-383-0
:GHS07
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:175.0 to 180.0 deg-C
Specification Test Results
Frequently Bought Together