Dimethyl adipate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:627-93-0
:
Formula:C₈H₁₄O₄
MDL Number:MFCD00008469
:174.2 g/mol
Merck Index:14,00162
Boiling Pt:~ 229 °C (1013 hPa)
Melting Pt:10.3 °C
Density:1,06 g/cm³ (20 °C)
Flash Pt:122°C
Storage Temperature:Ambient
EINECS:211-020-6
Specification Test Results
- Appearance:Colorless to Almost colorlessclear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together