Order Entry
Italy
ContactUsLinkComponent
Adipic acid ≥99.0% (by GC, titration analysis)
Irritant
Adipic acid ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Adipic acid ≥99.0% (by GC, titration analysis)
Supplier:  TCI
Certificates

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    152.0 to 155.0 deg-C
  • Solubility in Methanol:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together