Order Entry
Italy
ContactUsLinkComponent
 
 
 
1,6-Diaminohexane ≥99.0% (by GC, titration analysis)
 :  TCI
 
undefined
 
 
1,6-Diaminohexane ≥99.0% (by GC, titration analysis)
 :  TCI

 

  • Appearance:
    White to Almost white powder to lump
  • Purity (GC):
    min. 99.0 %
  • Purity (Neutralization titration):
    min. 99.0 %
  • Melting point:
    40.0 to 44.0 deg-C

 

 

Frequently Bought Together