
Glutaric acid ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:110-94-1
:
Formula:C₅H₈O₄
MDL Number:MFCD00004410
:132.12 g/mol
Merck Index:14,04473
Boiling Pt:200°C
Melting Pt:98 °C
Density:1,43 g/cm³ (15 °C)
EINECS:203-817-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:96.0 to 100.0 deg-C
- Solubility in Water:almost transparency
Specification Test Results
Frequently Bought Together