
1,5-Dihydroxynaphthalene ≥98.0% (by GC)
: TCI
:83-56-7
:
:C₁₀H₈O₂
:MFCD00003980
:160.17 g/mol
:375 °C (1013 hPa)
:180 °C
:1,41 g/cm³ (20 °C)
:Ambient
:201-487-4
:9,III
:GHS07
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 98.0 %
Frequently Bought Together