
2,5-Difluorobenzophenone ≥96.0% (by GC)
Catalog # TCIAD3598-5G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange clear liquid
- Purity (GC):min. 96.0 %
Specification Test Results
About this item
:
:218.2 g/mol
Boiling Pt:123°C
EINECS:285-298-2
:GHS07
CAS Number:85068-36-6
Formula:C₁₃H₈F₂O
MDL Number:MFCD00009899