


1,4-Diaminobutane ≥98.0% (by GC, titration analysis)
: TCI
:110-60-1
:
:C₄H₁₂N₂
:MFCD00008235
:88.15 g/mol
:14,07947
:159°C
:25…28 °C
:0,88 g/cm³ (20 °C)
:60°C
:Refrigerator
:203-782-3
:2925
:4.1,II
:GHS02
:GHS05
:GHS06
- Appearance:White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Solubility in Water:almost transparency
Frequently Bought Together