Diethyl succinate ≥99.0% (by GC)
: TCI
: Succinic acid diethyl ester, 1,4-Butanedioic acid diethyl ester
:123-25-1
:
:C₈H₁₄O₄
:MFCD00009208
:196°C
:174.2 g/mol
:-19
:104°C
:12,09038
:Ambient
:204-612-0
- Environmentally Preferable:
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Frequently Bought Together