

2,4-Dichloro-5-nitrobenzotrifluoride ≥98.0% (by GC)
Catalog # TCIAD2213-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Light yellow to Yellow to Orange powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:54.0 to 56.0 deg-C
Specification Test Results
About this item
:
:260 g/mol
Melting Pt:59.5 °C
:GHS07
:GHS08
CAS Number:400-70-4
Formula:C₇H₂Cl₂F₃NO₂
MDL Number:MFCD00007073