


1,3-Diaminopropane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:109-76-2
:
Formula:C₃H₁₀N₂
MDL Number:MFCD00008228
:74.13 g/mol
Boiling Pt:133°C
Melting Pt:–12 °C
Density:0,863 g/cm³ (20 °C)
Flash Pt:49°C
Storage Temperature:Ambient
EINECS:203-702-7
UN:2734
ADR:8,II
:GHS02
:GHS05
:GHS06
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together