


2,4-Diaminotoluene ≥98.0% (by GC, titration analysis)
Catalog # TCIAD0123-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:White to Gray to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
- Melting point:97.0 to 100.0 deg-C
Specification Test Results
About this item
:
:122.17 g/mol
Boiling Pt:292°C
Melting Pt:99 °C
Density:1,045 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-453-1
UN:1709
ADR:6.1,III
:GHS06
:GHS08
:GHS09
CAS Number:95-80-7
Formula:C₇H₁₀N₂
MDL Number:MFCD00007804