Order Entry
Italy
ContactUsLinkComponent
1,2-Epoxytetradecane ≥95.0% (by GC)
Irritant
1,2-Epoxytetradecane ≥95.0% (by GC)
Supplier:  TCI
Certificates
undefined
Irritant
1,2-Epoxytetradecane ≥95.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

Specification Test Results

Related Information

Frequently Bought Together