Order Entry
Italy
ContactUsLinkComponent
1,2-Epoxyoctan ≥96.0% (by GC)
Flammable
Irritant
1,2-Epoxyoctan ≥96.0% (by GC)
Catalog # TCIAE0206-25ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
1,2-Epoxyoctan ≥96.0% (by GC)
Catalog # TCIAE0206-25ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

Specification Test Results

About this item

 :
 :128.21 g/mol
Boiling Pt:167°C
Density:0,8395 g/cm³ (0 °C)
Flash Pt:37°C
Storage Temperature:Refrigerator
EINECS:221-047-5
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:2984-50-1
Formula:C₈H₁₆O
MDL Number:MFCD00005157