


1,2-Propylenediamine ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:78-90-0
:
Formula:C₃H₁₀N₂
MDL Number:MFCD00008089
:74.13 g/mol
Merck Index:14,07852
Boiling Pt:120°C
Melting Pt:–37 °C
Density:0,8584 g/cm³ (20 °C)
Flash Pt:33°C
Storage Temperature:Ambient
EINECS:201-155-9
UN:2258
ADR:8,II
:GHS02
:GHS05
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
Specification Test Results
Frequently Bought Together