EDTA disodium salt dihydrate
Catalog # RC-048
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:372.24 g/mol
Merck Index:12,03556
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037