Order Entry
Italy
ContactUsLinkComponent
α,α,α,2,3,5,6-Heptafluorotoluene ≥97.0% (by GC)
Flammable
Irritant
α,α,α,2,3,5,6-Heptafluorotoluene ≥97.0% (by GC)
Catalog # TCIAT1854-5G
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
α,α,α,2,3,5,6-Heptafluorotoluene ≥97.0% (by GC)
Catalog # TCIAT1854-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

Specification Test Results

About this item

 :
 :218.07 g/mol
Boiling Pt:111 - 112 °C (1013 hPa)
Density:1,601 g/cm³ (20 °C)
Flash Pt:32°C
UN:1993
ADR:3,III
 :GHS02
 :GHS07
Storage Temperature:Refrigerator
CAS Number:651-80-9
Formula:C₇HF₇
MDL Number:MFCD00012385