
1,16-Hexadecanedioic acid ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:505-54-4
:
Formula:C₁₆H₃₀O₄
MDL Number:MFCD00002746
Boiling Pt:430 °C (1013 hPa)
Melting Pt:122…123 °C
:286.41 g/mol
Storage Temperature:Ambient
EINECS:208-013-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:124.0 to 128.0 deg-C
- Solubility in hot Methanol:almost transparency
Specification Test Results
Frequently Bought Together