
1,13-Tridecanedioic acid ≥96.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:505-52-2
:
Formula:C₁₃H₂₄O₄
MDL Number:MFCD00002740
:244.33 g/mol
Boiling Pt:230 °C (0,5 -rr)
Melting Pt:108…111 °C
Density:1,15 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:208-011-4
:GHS07
Specification Test Results
- Appearance:White to Light yellow to Dark green powder to crystal
- Purity (GC):min. 96.0 %
- Purity (Neutralization titration):min. 96.0 %
- Melting point:112.0 to 116.0 deg-C
Specification Test Results
Frequently Bought Together