

(S)-(-)-2-Chloropropionic acid ≥98.0% (by GC, titration analysis)
: TCI
:29617-66-1
:
:H₃CCHClCOOH
:MFCD00064205
:108.52 g/mol
:186 °C (1013 hPa)
:5 °C
:1,255 g/cm³ (20 °C)
:Ambient
:411-150-5
:2511
:8,III
:GHS05
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 98.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:-12.0 to -16.0 deg (neat)
Frequently Bought Together