
(4-Fluorophenyl)acetic acid ≥98.0% (by titrimetric analysis)
: TCI
:405-50-5
:
:(C₆H₄F)CH₂COOH
:MFCD00004343
:154.14 g/mol
:13,04204
:164°C
:84…85 °C
:206-972-4
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (Neutralization titration):min. 98.0 %
- Melting point:83.0 to 86.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together