
(4-Chlorophenyl)acetic acid ≥97.0% (by titrimetric analysis)
: TCI
:1878-66-6
:
:(C₆H₄Cl)CH₂COOH
:MFCD00004344
:170.6 g/mol
:292 °C (1013 hPa)
:101…104 °C
:1,43 g/cm³ (20 °C)
:Ambient
:217-521-6
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 95.0 %
- Purity (Neutralization titration):min. 97.0 %
- Melting point:105.0 to 108.0 deg-C
- Solubility in Methanol:almost transparency
Frequently Bought Together