
(3-Chlorophenyl)acetic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:1878-65-5
:
Formula:(C₆H₄Cl)CH₂COOH
MDL Number:MFCD00004332
:170.6 g/mol
Boiling Pt:292 °C (1013 hPa)
Melting Pt:78…79 °C
Storage Temperature:Ambient
EINECS:217-520-0
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:77.0 to 81.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together