D-(-)-Sorbitol ≥97.0% (by GC)
: TCI
: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
:50-70-4
:98201-93-5
:HOH₂C(CH(OH))₄CH₂OH
:MFCD00004708
:200°C
:182.17 g/mol
:14,08725
:Ambient
:200-061-5
- Environmentally Preferable:
- Appearance:White powder to crystal
- Purity (GC):min. 97.0 %
- Specific rotation [a]20/D:+102 to +110 deg (C=0.4, Acidified (NH4)6Mo7O24)
- Solubility in Water:almost transparency
Frequently Bought Together